missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Janus Green B, pure, high purity biological stain
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Dye Content | 50% min. |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Merck Index | 15, 5303 |
| Color | Black or Brown/Green |
| Solubility Information | Solubility in water: soluble. |
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 511.07 |
| Grade | Pure |
| Show More |
RUO – Research Use Only