missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Janus Green B, pure, high purity biological stain

Catalog No. AC191680050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191680050 5 g Glass bottle
AC191680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191680050 Supplier Thermo Scientific Chemicals Supplier No. 191680050
Only null left

CAS: 2869-83-2 | C30H31ClN6 | 511.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2869-83-2
Molecular Formula C30H31ClN6
Molecular Weight (g/mol) 511.07
MDL Number MFCD00011758
InChI Key XXACTDWGHQXLGW-UHFFFAOYSA-M
Synonym C.I. 11050
PubChem CID 76123
ChEBI CHEBI:21184
IUPAC Name 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
SMILES [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C

Specifications

Dye Content 50% min.
Merck Index 15, 5303
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 511.07
Grade Pure
Packaging Glass bottle
Color Black or Brown/Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Janus Green B, Pure
Show More Show Less

RUO – Research Use Only