Learn More
Thermo Scientific Chemicals Janus Green B
CAS: 2869-83-2 | C30H31ClN6 | 511.07 g/mol
$118.63 - $372.48
Chemical Identifiers
| CAS | 2869-83-2 |
|---|---|
| Molecular Formula | C30H31ClN6 |
| Molecular Weight (g/mol) | 511.07 |
| MDL Number | MFCD00011758 |
| InChI Key | XXACTDWGHQXLGW-UHFFFAOYSA-M |
| Synonym | C.I. 11050 |
| PubChem CID | 76123 |
| ChEBI | CHEBI:21184 |
| IUPAC Name | 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride |
| SMILES | [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1739106
|
Thermo Scientific Chemicals
A1739106 |
5 g |
Each for $118.63
|
|
|||||
|
AAA1739114
|
Thermo Scientific Chemicals
A1739114 |
25 g |
Each for $372.48
|
|
|||||
Description
Janus Green B is a stain that interacts with DNA and has been used for histology studies including mitochondrial staining, in which its oxidation and reduction reveals alterations to the electron transfer chain. The stain has been utilized in studies for effective and rapid staining of insect chordotonal organs and peripheral nerves, where the nerves appear dark blue in comparison to non stained tissue. With the use of Janus Green B, rabbit lymphatic vessel studies demonstrate that threadlike structures within the lymphatic vessels, known as Bonghan ducts, contain a high density of mitochondria in their cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsJanus Green B is a stain that interacts with DNA and has been used for histology studies including mitochondrial staining, in which its oxidation and reduction reveals alterations to the electron transfer chain. The stain has been utilized in studies for effective and rapid staining of insect chordotonal organs and peripheral nerves, where the nerves appear dark blue in comparison to non stained tissue. With the use of Janus Green B, rabbit lymphatic vessel studies demonstrate that threadlike structures within the lymphatic vessels, known as Bonghan ducts, contain a high density of mitochondria in their cells.
Solubility
Soluble in water (20 mg/ml at 20°C), ethanol, and slightly soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 2869-83-2 | |
| 511.07 | |
| XXACTDWGHQXLGW-UHFFFAOYSA-M | |
| 76123 | |
| 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride |
| C30H31ClN6 | |
| MFCD00011758 | |
| C.I. 11050 | |
| CHEBI:21184 | |
| [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C |
Specifications
| 2869-83-2 | |
| MFCD00011758 | |
| C.I. 11050 | |
| XXACTDWGHQXLGW-UHFFFAOYSA-M | |
| 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride | |
| 76123 | |
| 511.07 | |
| Janus Green B |
| C30H31ClN6 | |
| 14,5255 | |
| Soluble in water (20mg/ml at 20°C),ethanol,and slightly soluble in alcohol. | |
| [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C | |
| 511.07 | |
| CHEBI:21184 | |
| 5 g |
Safety and Handling
EINECSNumber : 220-695-6
RTECSNumber : SG1633000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only