Learn More
Ansell KleenGuard™ V20 Purity™ Safety Glasses
GREENER_CHOICE
Catalog No. 19170101
Change view
Click to view available options
Lens Tint:
Clear
Lens Coating:
Anti-fog
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19170101 Clear Uncoated
19413028 Clear Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19170101 Supplier Ansell Supplier No. 25650
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KleenGuard Purity Safety Eyewear allows you to provide stylish, yet economical eye protection for yourself and your team.
  • Comfortable, padded temples for enhanced comfort
  • Clear lenses allow maximum visible light transmission; ideal for indoor use
  • Anti-Fog coating to prevent fogging; ideal for humid environments and temperature variances
  • Hardcoated lenses for scratch resistance and longer life
  • Lenses are made of a polycarbonate material that provides 99.9% UVA/UVB/UVC protection
  • Meets ANSI Z87.1+ standards for high impact (25654-00 meets ANSI Z87.1+2010 and 25654-01 meets ANSI Z87.1+2015) and certified to meet the requirements of CSA Z94.3
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1: 2010; EN 166: 2001; CSA Z94.3:2007; AS/NZS 1337.1:2010
Lens Tint Clear
For Use With (Application) Industrial manufacturing, outdoor environments, impact, splash, grinding/chipping, shaving/sanding
Frame Color Clear
Temple Style Ratcheted
Temple Color Black
Lens Material Polycarbonate
Frame Material Polycarbonate