missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAC Medical Phlebotomy Lab Trays
Mini lab tray is single-purpose use
$251.00 - $255.57
Specifications
| Color | White |
|---|---|
| Material | Polystyrene |
| Product Type | Phlebotomy Lab Tray |
| For Use With (Application) | For single-purpose |
Description
- Green high-impact polystyrene
- Large lab tray carries up to 80 tubes and many other components
- Compact lab tray has a medium capacity for up to 60 tubes and selected components
Order Info
Handles sold separately: 22-038-419 (JAC 8); 22-038-420 (JAC 9). Lid not included.
Specifications
| White | |
| Polystyrene | |
| Phlebotomy Lab Tray | |
| For single-purpose |