missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine Hydrochloride 98.0+%, TCI America™

Catalog No. I08471G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08471G 1 g
I0847200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08471G Supplier TCI America Supplier No. I08471G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148849-67-6
Molecular Formula C27H37ClN2O5
Molecular Weight (g/mol) 505.052
MDL Number MFCD00929899
InChI Key HLUKNZUABFFNQS-ZMBIFBSDSA-N
PubChem CID 3045381
ChEBI CHEBI:85969
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-methylamino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
SMILES CN(CCCN1CCC2=CC(=C(C=C2CC1=O)OC)OC)CC3CC4=CC(=C(C=C34)OC)OC.Cl

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 505.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ivabradine Hydrochloride
TSCA No
Recommended Storage Refrigerator