Learn More
Biogems International Inc ITS (Insulin-Transferrin-Selenium) (100x) 10mL
Catalog No. 501528630
Encompass
Insulin-Transferrin-Selenium (ITS) is a basal media supplement and can be used to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Insulin promotes the uptake of glucose and amino acids, transferrin is a non-toxic iron carrier, selenium is required for the function of glutathione peroxidase and other antioxidant enzymes. The components of the solution are manufactured animal-free and it is provided as a 100x sterile concentrate.
Catalog No. 501528630 Supplier Biogems International Inc Supplier No. 0010210
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Insulin-Transferrin-Selenium (ITS) is a basal media supplement and can be used to reduce the amount of fetal bovine serum (FBS) needed to culture cells. Insulin promotes the uptake of glucose and amino acids, transferrin is a non-toxic iron carrier, selenium is required for the function of glutathione peroxidase and other antioxidant enzymes. The components of the solution are manufactured animal-free and it is provided as a 100x sterile concentrate.