Learn More
Pyramex™ Itek™ Safety Eyewear
Description
Specifications
Specifications
| Lens Coating | Uncoated |
| Certifications/Compliance | ANSI Z87.1, CSA Z94.3-07 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Blue |
| Weight (Metric) | 23.5 g |
| Lens Material | Polycarbonate |