Learn More
Pyramex™ Itek™ Safety Eyewear
Lightweight, frameless protection is ideal for all-day wear
Supplier: Pyramex™ S5810ST
Description
Specifications
| H2X Anti-fog | |
| EN-166 | |
| 23.5 g |
| ANSI Z87.1, CSA Z94.3-07 | |
| Clear | |
| Polycarbonate |