missing translation for 'onlineSavingsMsg'
Learn More

Itaconic Anhydride 95.0+%, TCI America™

Supplier:  TCI America I0203500G

 View more versions of this product

Catalog No. I0203500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Itaconic Anhydride
2170-03-8
115°C
Crystal-Powder at 20°C
MFCD00005530
OFNISBHGPNMTMS-UHFFFAOYSA-N
3-methylideneoxolane-2,5-dione
75110
≥95.0% (T)
68°C
White-Yellow
500 g
C5H4O3
itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
C=C1CC(=O)OC1=O
112.08
112.08

Chemical Identifiers

2170-03-8
112.08
OFNISBHGPNMTMS-UHFFFAOYSA-N
75110
C=C1CC(=O)OC1=O
C5H4O3
MFCD00005530
itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
3-methylideneoxolane-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1902

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions