missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Itaconic anhydride, 97%
Description
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Specifications
Specifications
| Melting Point | 68°C to 72°C |
| Quantity | 25 g |
| Beilstein | 2212 |
| Boiling Point | 114°C to 115°C (12 mmHg) |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 112.09 |
| Percent Purity | 97% |
| Chemical Name or Material | Itaconic anhydride |
Safety and Handling
| missing translation for 'healthHazard1' | H318 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P280-P305+P351+P338-P310 |
| EINECSNumber | 218-518-2 |
| TSCA | Yes |
| Recommended Storage | Store under Nitrogen |
RUO – Research Use Only