missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carboxylic acid, 95%, Thermo Scientific™

$114.99 - $114.99

Chemical Identifiers

CAS 21169-71-1
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD00156151
InChI Key MIIQJAUWHSUTIT-UHFFFAOYSA-N
Synonym isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
PubChem CID 2060599
IUPAC Name 1,2-oxazole-5-carboxylic acid
SMILES C1=C(ON=C1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC10301DE
SDS View Documents
Thermo Scientific Chemicals
CC10301DE
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

21169-71-1
113.072
MIIQJAUWHSUTIT-UHFFFAOYSA-N
2060599
C1=C(ON=C1)C(=O)O
C4H3NO3
MFCD00156151
isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
1,2-oxazole-5-carboxylic acid

Specifications

21169-71-1
100.0
C4H3NO3
5 g
MIIQJAUWHSUTIT-UHFFFAOYSA-N
1,2-oxazole-5-carboxylic acid
2060599
97%
96.5
97%
MFCD00156151
isoxazole-5-carboxylic acid, 5-isoxazolecarboxylic acid, isoxazole-5-carboxylicacid, 5-carboxyisoxazole, pubchem8643, isoxazole-5-carboxylic, 5-carboxy-1,2-oxazole, isoxazole 5-carboxylic acid, ksc207s2h
C1=C(ON=C1)C(=O)O
113.072
113.07
Isoxazole-5-carboxylic acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'