missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™

Catalog No. CC10302DA
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC10302DA 1g
CC10302EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC10302DA Supplier Thermo Scientific Chemicals Supplier No. CC10302DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62348-13-4
Molecular Formula C4H2ClNO2
Molecular Weight (g/mol) 131.52
MDL Number MFCD00067868
InChI Key NASLINFISOTVJJ-UHFFFAOYSA-N
Synonym isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, isoxazole-5-carbonylchloride, isoxazole-5-carboxylic acid chloride, acmc-1bhwq, 5-isoxazolecarbonylchloride, isoxazol-5-carbonyl chloride, isoxazole-5-carbonyl-chloride, 5-isoxazolecarbonyl chloride 9ci
PubChem CID 2736707
IUPAC Name 1,2-oxazole-5-carbonyl chloride
SMILES ClC(=O)C1=CC=NO1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 131.52
Percent Purity 90%
Chemical Name or Material Isoxazole-5-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.