missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbonyl chloride, 97%

Catalog No. AAA1462506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1462506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1462506 Supplier Thermo Scientific Chemicals Supplier No. A1462506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62348-13-4 | C4H2ClNO2 | 131.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62348-13-4
Molecular Formula C4H2ClNO2
Molecular Weight (g/mol) 131.52
MDL Number MFCD00067868
InChI Key NASLINFISOTVJJ-UHFFFAOYSA-N
Synonym isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, isoxazole-5-carbonylchloride, isoxazole-5-carboxylic acid chloride, acmc-1bhwq, 5-isoxazolecarbonylchloride, isoxazol-5-carbonyl chloride, isoxazole-5-carbonyl-chloride, 5-isoxazolecarbonyl chloride 9ci
PubChem CID 2736707
IUPAC Name 1,2-oxazole-5-carbonyl chloride
SMILES ClC(=O)C1=CC=NO1

Specifications

Density 1.362
Boiling Point 167°C to 168°C
Refractive Index 1.499
Quantity 5 g
UN Number UN3265
Beilstein 4375023
Sensitivity Moisture sensitive
Formula Weight 131.52
Percent Purity 97%
Chemical Name or Material Isoxazole-5-carbonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only