missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-3-carboxylic acid, 97%

Catalog No. AAH5843806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5843806 5 g
AAH5843803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5843806 Supplier Thermo Scientific Chemicals Supplier No. H5843806

missing translation for 'validMainframeMsg'

CAS: 3209-71-0 | C4H3NO3 | 113.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3209-71-0
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.07
MDL Number MFCD06738796
InChI Key UXYRXGFUANQKTA-UHFFFAOYSA-N
Synonym 3-isoxazolecarboxylic acid, isoxazole-3-carboxylic acid, isoxazole-3-carboxylicacid, 3-isoxazolecarboxylicacid, 3-carboxyisoxazole, isoxazole-3-carboxylic, pubchem12341, acmc-209hqw, ksc222a3r, aronis24546
PubChem CID 11286453
IUPAC Name 1,2-oxazole-3-carboxylic acid
SMILES OC(=O)C1=NOC=C1

Specifications

Quantity 5 g
Beilstein 110919
Formula Weight 113.07
Percent Purity 97%
Chemical Name or Material Isoxazole-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only