missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl Chloride 99.0+%, TCI America™

Catalog No. I0200500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0200500ML 500 mL
I020025ML 25 mL
I0200100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0200500ML Supplier TCI America Supplier No. I0200500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl

Specifications

Color Yellow
Boiling Point 116°C
Quantity 500 mL
UN Number 2502
Formula Weight 120.58
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Isovaleryl Chloride
EINECSNumber (2)-0632
TSCA Yes