missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 98%

CAS: 108-12-3 | C5H9ClO | 120.58 g/mol

$67.84 - $67.84

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169280250
SDS View Documents
Thermo Scientific Chemicals
169280250
25 g Glass bottle
Each for $67.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-12-3
120.58
ISULZYQDGYXDFW-UHFFFAOYSA-N
66054
CC(C)CC(=O)Cl
C5H9ClO
MFCD00000738
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
3-methylbutanoyl chloride

Specifications

108-12-3
0.9800g/mL
27°C
98%
C5H9ClO
(CH3)2CHCH2COCl
25 g
0.98
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
ISULZYQDGYXDFW-UHFFFAOYSA-N
3-methylbutanoyl chloride
66054
98%
Isovaleryl chloride, 98%
Colorless to Yellow or Pink
115°C to 117°C
Authentic
Glass bottle
1.4150 to 1.417
MFCD00000738
02, 315
15, 5278
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
CC(C)CC(=O)Cl
120.58
120.58
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-552-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only