missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 97%

CAS: 108-12-3 | C5H9ClO | 120.576 g/mol

Supplier:  Thermo Scientific Chemicals B2400222

 View more versions of this product

Catalog No. AAB2400222


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isovaleryl chloride
0.989
27°C (80°F)
1.416
100 g
741910
14,5232
ISULZYQDGYXDFW-UHFFFAOYSA-N
3-methylbutanoyl chloride
66054
98%
108-12-3
115°C to 117°C
C5H9ClO
MFCD00000738
UN2920
Moisture sensitive
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
CC(C)CC(=O)Cl
120.576
120.58

Chemical Identifiers

108-12-3
120.576
ISULZYQDGYXDFW-UHFFFAOYSA-N
66054
CC(C)CC(=O)Cl
C5H9ClO
MFCD00000738
isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
3-methylbutanoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P374-P380-P501c

H226-H290-H314-H331

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 203-552-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only