missing translation for 'onlineSavingsMsg'
Learn More

Isovaleryl chloride, 97%

Catalog No. AAB2400214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2400214 25 g
AAB2400222 100 g
AAB2400236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2400214 Supplier Thermo Scientific Chemicals Supplier No. B2400214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-12-3 | C5H9ClO | 120.576 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-12-3
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00000738
InChI Key ISULZYQDGYXDFW-UHFFFAOYSA-N
Synonym isovaleryl chloride, butanoyl chloride, 3-methyl, isovaleroyl chloride, isopentanoyl chloride, isovaleric acid chloride, 3-methylbutyryl chloride, isovalerylchloride, 3-methyl-butanoyl chloride, unii-j2ml32m57v, 3-methyl-butyryl chloride
PubChem CID 66054
IUPAC Name 3-methylbutanoyl chloride
SMILES CC(C)CC(=O)Cl

Specifications

Density 0.989
Boiling Point 115°C to 117°C
Flash Point 27°C (80°F)
Refractive Index 1.416
Quantity 25 g
UN Number UN2920
Beilstein 741910
Sensitivity Moisture sensitive
Merck Index 14,5232
Formula Weight 120.58
Percent Purity 98%
Chemical Name or Material Isovaleryl chloride
Show More Show Less
Hazard Category H226-H290-H314-H331
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P234-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 203-552-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only