Learn More
Isovaleric anhydride, 95%
CAS: 1468-39-9 | C10H18O3 | 186.25 g/mol
Supplier: Thermo Scientific Chemicals L0438514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isovaleric anhydride | |
| -65°C | |
| 103°C to 104°C (24 mmHg) | |
| C10H18O3 | |
| [(CH3)2CHCH2CO]2O | |
| 25 g | |
| 1635850 | |
| isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride | |
| CC(C)CC(=O)OC(=O)CC(C)C | |
| 186.25 | |
| 186.25 |
| 1468-39-9 | |
| 0.927 | |
| 91°C (195°F) | |
| 1.4165 | |
| MFCD00015051 | |
| UN3265 | |
| Moisture sensitive | |
| FREZLSIGWNCSOQ-UHFFFAOYSA-N | |
| 3-methylbutanoyl 3-methylbutanoate | |
| 73847 | |
| 95% |
Chemical Identifiers
| 1468-39-9 | |
| 186.25 | |
| FREZLSIGWNCSOQ-UHFFFAOYSA-N | |
| 73847 | |
| CC(C)CC(=O)OC(=O)CC(C)C |
| C10H18O3 | |
| MFCD00015051 | |
| isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride | |
| 3-methylbutanoyl 3-methylbutanoate |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P234-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H227-H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 215-994-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only