missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric anhydride, 95%

Catalog No. aal0438506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0438506 5 g
AAL0438514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0438506 Supplier Thermo Scientific Chemicals Supplier No. L0438506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1468-39-9 | C10H18O3 | 186.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1468-39-9
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00015051
InChI Key FREZLSIGWNCSOQ-UHFFFAOYSA-N
Synonym isovaleric anhydride, 3-methylbutanoic anhydride, butanoic acid, 3-methyl-, anhydride, butanoic acid, 3-methyl-, 1,1'-anhydride, isovalericanhydride, isovaleriansaureanhydrid, iso-pentanoic anhydride, acmc-209cxm, isovaleric anhydride gc, butanoic acid,3-methyl-,1,1'-anhydride
PubChem CID 73847
IUPAC Name 3-methylbutanoyl 3-methylbutanoate
SMILES CC(C)CC(=O)OC(=O)CC(C)C

Specifications

Melting Point -65°C
Density 0.927
Boiling Point 103°C to 104°C (24 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.4165
Linear Formula [(CH3)2CHCH2CO]2O
Quantity 5 g
UN Number UN3265
Beilstein 1635850
Sensitivity Moisture sensitive
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Isovaleric anhydride
Show More Show Less
Hazard Category H227-H290-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P234-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 215-994-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only