missing translation for 'onlineSavingsMsg'
Learn More

Isovaleric Acid 99.0+%, TCI America™

Catalog No. p-7132041
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M018225ML 25 mL
M0182500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M018225ML Supplier TCI America Supplier No. M018225ML
Only null left

Chemical Identifiers

CAS 503-74-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00002726
InChI Key GWYFCOCPABKNJV-UHFFFAOYSA-N
Synonym isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid
PubChem CID 10430
ChEBI CHEBI:28484
IUPAC Name 3-methylbutanoic acid
SMILES CC(C)CC(O)=O

Specifications

Color Colorless
Boiling Point 175°C
Quantity 25 mL
UN Number 2922
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isovaleric Acid
EINECSNumber (2)-0608
RTECSNumber NY1400000
TSCA Yes