missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide isovalérique 99,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 175°C |
| Quantity | 25 mL |
| UN Number | 2922 |
| Formula Weight | 102.13 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Isovaleric Acid |
Sécurité et manipulation
| EINECSNumber | (2)-0608 |
| RTECSNumber | NY1400000 |
| TSCA | Yes |