Learn More
Isovaleric acid, 98%
CAS: 503-74-2 | C5H10O2 | 102.13 g/mol
Supplier: Thermo Scientific Chemicals A18642AU
Description
A molecular entity capable of donating a hydron to an acceptor (Bronsted base). It is a favorable carbon source for cell growth. Moreover, it is a promising odor indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA molecular entity capable of donating a hydron to an acceptor (Bronsted base). It is a favorable carbon source for cell growth. Moreover, it is a promising odor indicator.
Solubility
Soluble in water 20°C 25 g/L. Soluble in ethanol, chloroform, ether.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Isovaleric acid | |
| -33°C | |
| 3 | |
| 70°C (158°F) | |
| Rancid | |
| 1.403 | |
| 1000 mL | |
| 1098522 | |
| isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid | |
| GWYFCOCPABKNJV-UHFFFAOYSA-N | |
| 3-methylbutanoic acid | |
| 10430 | |
| 102.13 | |
| Liquid |
| 503-74-2 | |
| 0.937 | |
| 175°C to 177°C | |
| 0.98 | |
| C5H10O2 | |
| MFCD00002726 | |
| UN3265 | |
| 14,5231 | |
| Soluble in water 20°C 25g/L. Soluble in ethanol,chloroform,ether. | |
| CC(C)CC(O)=O | |
| 102.13 | |
| CHEBI:28484 | |
| 98% |
Chemical Identifiers
| 503-74-2 | |
| 102.13 | |
| GWYFCOCPABKNJV-UHFFFAOYSA-N | |
| 10430 | |
| 3-methylbutanoic acid |
| C5H10O2 | |
| MFCD00002726 | |
| isovaleric acid, isopentanoic acid, 3-methylbutyric acid, delphinic acid, isopropylacetic acid, butanoic acid, 3-methyl, isovalerianic acid, isobutylformic acid, 3-methylbutyrate, beta-methylbutyric acid | |
| CHEBI:28484 | |
| CC(C)CC(O)=O |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H311-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 207-975-3
RTECSNumber : NY1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only