missing translation for 'onlineSavingsMsg'
Learn More

Isovaleramide, 98%

Catalog No. AAL0907206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0907206 5 g
AAL0907214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0907206 Supplier Thermo Scientific Chemicals Supplier No. L0907206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 541-46-8 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-46-8
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014807
InChI Key SANOUVWGPVYVAV-UHFFFAOYSA-N
Synonym isovaleramide, butanamide, 3-methyl, 3-methylbutyramide, isovaleric amide, isopentanamide, beta-methylbutyramide, isovaleric acid amide, unii-9cp4kb634m, isovaleramide usan, .beta.-methylbutyramide
PubChem CID 10930
IUPAC Name 3-methylbutanamide
SMILES CC(C)CC(=O)N

Specifications

Melting Point 134°C to 138°C
Boiling Point 232°C
Quantity 5 g
Beilstein 1740789
Merck Index 14,5230
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material Isovaleramide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-781-1
RTECSNumber EJ3650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only