missing translation for 'onlineSavingsMsg'
Learn More

Isovaleraldehyde 98.0+%, TCI America™

Catalog No. I0192500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0192500ML 500 mL
I019225ML 25 mL
I0192100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0192500ML Supplier TCI America Supplier No. I0192500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 590-86-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007014
InChI Key YGHRJJRRZDOVPD-UHFFFAOYSA-N
Synonym isovaleraldehyde, 3-methylbutyraldehyde, isopentaldehyde, isovaleral, isovalerylaldehyde, isoamylaldehyde, butanal, 3-methyl, isopentanal, beta-methylbutanal, isovaleric aldehyde
PubChem CID 11552
ChEBI CHEBI:16638
IUPAC Name 3-methylbutanal
SMILES CC(C)CC=O

Specifications

Color Yellow
Boiling Point 93°C
Quantity 500 mL
UN Number 2058
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isovaleraldehyde
EINECSNumber (2)-0494
RTECSNumber ES3450000
TSCA Yes