missing translation for 'onlineSavingsMsg'
Learn More

Isovaleraldehyde, 98%

Catalog No. aaa19640ae
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
1000 mL
500 mL
Catalog No. Quantity
AAA19640AE 100 mL
AAA19640AP 500 mL
AAA19640AU 1000 mL
3 options

Catalog No. AAA19640AE

Supplier: Thermo Scientific Chemicals A19640AE

Only null left

CAS: 590-86-3 | C5H10O | 86.13 g/mol

Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isovaleraldehyde acts as a reagent in the preparation of active pharmaceutical ingredient (API) products. It serves as an internal standard for the determination of wine aroma carbonyl compounds with 5-8 carbon atoms. Further, it is utilized as a standard to evaluate the quality of olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. In addition to this, it is employed to enhance the taste and odor of the food.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, alkaline materials and reducing agents. Store in a cool place.

Chemical Identifiers

CAS 590-86-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007014
InChI Key YGHRJJRRZDOVPD-UHFFFAOYSA-N
Synonym isovaleraldehyde, 3-methylbutyraldehyde, isopentaldehyde, isovaleral, isovalerylaldehyde, isoamylaldehyde, butanal, 3-methyl, isopentanal, beta-methylbutanal, isovaleric aldehyde
PubChem CID 11552
ChEBI CHEBI:16638
IUPAC Name 3-methylbutanal
SMILES CC(C)CC=O

Specifications

Melting Point -51°C
Density 0.8
Boiling Point 91°C to 93°C
Flash Point −1°C (30°F)
Odor Fruit-like
Refractive Index 1.3882
Quantity 100 mL
UN Number UN2058
Beilstein 773692
Sensitivity Air Sensitive
Merck Index 14,5229
Solubility Information Miscible with alcohol and diethyl ether. Slightly miscible with water.
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material Isovaleraldehyde
Show More Show Less
Hazard Category H225-H317-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VALERALDEHYDE
EINECSNumber 209-691-5
RTECSNumber ES3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only