missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isotetrandrine, 98%

Catalog No. AAJ67081MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67081MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67081MCR Supplier Thermo Scientific Chemicals Supplier No. J67081MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 477-57-6
Molecular Formula C38H42N2O5
Synonym Berbamine methyl ether; 6,6',7,12-Tetramethoxy-2,2'-dimethylberbaman

Specifications

Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
Formula Weight 606.76
Product Type Isotetrandrine, 98%
Form Solid
Assay 98%

RUO – Research Use Only