missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Value Lab Refrigerator/Freezer

Catalog No. FBV05CPSA
Change view
Click to view available options
Capacity (English) Refrigerator:
5.1 cu. ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Refrigerator
FBV05CPSA 5.1 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FBV05CPSA Supplier Fisherbrand™ Supplier No. FBV05CPSA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill, freeze and retrieve samples safely, simply, and economically.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish

Specifications

Type General Purpose Combo
Capacity (English) Refrigerator 5.1 cu. ft.
Capacity (Metric) Refrigerator 144.4 L
Temperature Range (Freezer) -12°C to -20°C
Temperature Range (Refrigerator) 1°C to 12°C
Temperature (Setpoint) Adjustable
Door Style Hinged, Open Left to Right
No. of Doors 1
Inner Doors 1
Shelves 2 Adjustable
Door Shelves 2
Compressor Hermetic
Defrost Automatic (Refrigerator), Manual (Freezer)
Access Security None
Refrigerant R600a hydrocarbon refrigerant
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor General Storage of Non-Critical Samples and Reagents
Min. Clearance 4 in. on all sides
Typical Lifetime 10 Years
Warranty 12 Months
Dimensions (DxWxH) Exterior 23.5 x 23.63 x 33.5 in. (56.69 x 60.02 x 85.1 cm)
Dimensions (D x W x H) Interior Refrigerator: 17 x 20.5 x 19.5 in.(43.2 x 52.1 x 49.5 cm); Freezer: 11.5 x 19 x 5 in. (29.2 x 48.3 x 12.7 cm)
Energy Usage 0.35 kWh/Day
Amperage 1.2 A
Plug Type NEMA 5-15 P
Voltage 115 V
Frequency 60 Hz
Shipping Weight (Metric) 46 kg
Show More Show Less