Learn More
Fisherbrand™ Isotemp Series General Purpose Ultra-Low Freezer
Catalog No. FLNPLC
Change view
Click to view available options
Type:
CO2 Backup System
LN2 Backup System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
FLNPLC LN2 Backup System
FCOPLC CO2 Backup System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FLNPLC Supplier Fisherbrand™ Supplier No. FLNPLC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Isotemp™ ULT Freezers combine reliability and performance with cost-effective operation and innovative features.
  • Isotemp™ Ultra-Low-Temperature Freezers combine reliability and performance with cost-effective operation and innovative features.
  • They are built using an advanced technology platform, refrigeration, microprocessor controls, and high-quality construction.
  • Performance
  • Maximum temperature recovery: Reduced/low peak temperature variation and fast open-door recovery
  • More comfortable lab environment: Low noise levels (like your refrigerator at home2)
  • Operational efficiency: Lower energy consumption and HVAC costs
  • User Friendly
  • Single-handed door operation
  • Five inner compartments (with doors)
  • Large temperature display with touch control
  • Centralized settings and controls with battery backup
  • Alarms, security passcode, and padlock-compatible doors
  • Easy access to the filter/grill assembly (cleaning and maintenance)
  • Sustainability
  • Manufactured in award winning zero-waste to landfill facility
  • Natural, green hydrocarbon refrigerants improve performance and increase reliability with low GWP and zero ODP
  • Environmentally friendly: Water-blown foam insulation; compliant with EPA SNAP and EU F-Gas regulations
  • Additional features
  • Standard RS-485, 4 - 20 mA analog outputs and dry contacts included
  • Battery backup for control panel (power interruptions)
  • Four capacities; Universal voltage
  • No setup required
  • Warranty
  • Five years parts and labor
Specifications
Type LN2 Backup System
For Use With (Equipment) Isotemp Series Upright ULT Freezers
Includes Customer Installed LN2 Backup System