missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
$26,141.24 - $31,077.00
Specifications
| Temperature Range | 2°C to 10°C |
|---|---|
| Temperature (Setpoint) | 5°C |
| Refrigerant | R290 |
| Door Style | Glass |
| Capacity (English) Shelf | 100 lb. |
| Catalog Number | Mfr. No. | Capacity (English) | Shelves | Color | Interior | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Shelves | Color | Interior | Price | Quantity & Availability | ||
FBV72RPGA
![]() |
Fisherbrand™
FBV72RPGA |
72 cu. ft. | 12 Full | White | Painted White Steel |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
FBV72RSGA
![]() |
Fisherbrand™
FBV72RSGA |
72 cu. ft. | 12 Full | Gray | Stainless Steel |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
Specifications
| 2°C to 10°C | |
| R290 | |
| 100 lb. | |
| cULus, ENERGYSTAR | |
| 1 Rear access port | |
| NEMA 5-15 P |
| 5°C | |
| Glass | |
| 45 kg | |
| Microprocessor | |
| Key Lock | |
| 115 V 60 Hz |