missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators are designed with your needs in mind.

$21,126.40

Specifications

Temperature Range 2°C to 10°C
Temperature (Setpoint) 5°C
Refrigerant R290
Door Style Glass
Capacity (English) Shelf 100 lb./full shelf; 50 lb./half shelf
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (English) Shelves Color Interior Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Shelves Color Interior Price Quantity & Availability  
FBG72CPGA
Accessories View Documents Promotion Details
Fisherbrand™
FBG72CPGA
72 cu. ft. 12 Full, 2 Half White Painted White Steel N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Internal plug/outlet
  • Special safety sensor turns the internal plug/outlet off if a hydrocarbon (coolant) leak is detected
Specifications

Specifications

2°C to 10°C
R290
100 lb./full shelf; 50 lb./half shelf
cULus, ENERGYSTAR
1 Rear access port and 2 side access ports
Key Lock
115 V 60 Hz
5°C
Glass
45 kg/full shelf, 22.5 kg/half shelf
Microprocessor
2 years parts and labor, plus 3 year compressor warranty
NEMA 5-15 P
Videos
SDS
Documents

Documents