missing translation for 'onlineSavingsMsg'
Learn More

Isostearic Acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic Acid) 90.0+%, TCI America™

Catalog No. I018425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I018425G 25 g
I0184500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I018425G Supplier TCI America Supplier No. I018425G
Only null left

Chemical Identifiers

CAS 54680-48-7
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00135620
InChI Key QBTIHZIVENIGSW-UHFFFAOYSA-N
Synonym 2,2,4,8,10,10-Hexamethylundecane-5-carboxylic Acid, 2-(4,4-Dimethyl-2-pentyl)-5,7,7-trimethyl-n-octanoic Acid
PubChem CID 256370
IUPAC Name 2-(4,4-dimethylpentan-2-yl)-5,7,7-trimethyloctanoic acid
SMILES CC(CCC(C(C)CC(C)(C)C)C(=O)O)CC(C)(C)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 284.48
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Isostearic Acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic Acid)
EINECSNumber (2)-0608
TSCA Yes