missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Isosorbide | |
| 652-67-5 | |
| 175°C | |
| Crystal-Powder at 20°C | |
| MFCD00064827 | |
| KLDXJTOLSGUMSJ-JGWLITMVSA-N | |
| (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol | |
| 12597 | |
| ≥98.0% (GC) |
| 61°C | |
| White-Yellow | |
| 100 g | |
| C6H10O4 | |
| isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol | |
| C1C(C2C(O1)C(CO2)O)O | |
| 146.142 | |
| 146.14 |
Chemical Identifiers
| 652-67-5 | |
| 146.142 | |
| KLDXJTOLSGUMSJ-JGWLITMVSA-N | |
| 12597 | |
| C1C(C2C(O1)C(CO2)O)O |
| C6H10O4 | |
| MFCD00064827 | |
| isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol | |
| (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol |
Safety and Handling
RTECSNumber : LZ4380000
TSCA : Yes