missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide dimethyl ether, 98%

Catalog No. AAA1751622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1751622 100 g
AAA1751614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1751622 Supplier Thermo Scientific Chemicals Supplier No. A1751622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5306-85-4 | C8H14O4 | 174.20 g/mol

It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.

Solubility
Soluble in water.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.

Chemical Identifiers

CAS 5306-85-4
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00064828
InChI Key MEJYDZQQVZJMPP-UHFFFAOYNA-N
Synonym dimethyl isosorbide, isosorbide dimethyl ether, 3r,3ar,6s,6ar-3,6-dimethoxyhexahydrofuro 3,2-b furan, unii-sa6a6v432s, 1,4:3,6-dianhydro-2,5-di-o-methyl-d-glucitol, arlasolve dmi, 2,5-di-o-methyl-1,4:3,6-dianhydro-d-glucitol, dimethyl isosorbide ether, 3r,3ar,6s,6ar-3,6-dimethoxy-hexahydrofuro 3,2-b furan, 3s,3ar,6r,6ar-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro 3,2-b furan
PubChem CID 62990
IUPAC Name (3S,3aR,6R,6aR)-3,6-dimethoxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan
SMILES COC1COC2C(COC12)OC

Specifications

Melting Point -70°C
Density 1.165
Quantity 100 g
Refractive Index 1.462
Beilstein 80854
Boiling Point 93°C to 95°C (0.1 mmHg)
Solubility Information Soluble in water.
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material Isosorbide dimethyl ether
EINECSNumber 226-159-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only