missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide, 98%

Catalog No. p-3758266
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351910050 5 g Glass bottle
AC351911000 100 g Glass bottle
AC351915000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC351910050 Supplier Thermo Scientific Chemicals Supplier No. 351910050
Only null left

CAS: 652-67-5 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00064827
InChI Key KLDXJTOLSGUMSJ-JGWLITMVSA-N
Synonym isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol
PubChem CID 12597
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES C1C(C2C(O1)C(CO2)O)O

Specifications

Melting Point 56.0°C to 62.0°C
Color Beige or White to Yellow
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals, Crystalline Solid or Mass
Assay Percent Range 97.5% min. (GC)
Merck Index 15, 5270
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material Isosorbide
Show More Show Less
EINECSNumber 211-492-3

RUO – Research Use Only