missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoquinoline 95.0+%, TCI America™
$51.98 - $281.08
Chemical Identifiers
| CAS | 119-65-3 |
|---|---|
| Molecular Formula | C9H7N |
| Molecular Weight (g/mol) | 129.16 |
| MDL Number | MFCD00006898,MFCD31699977 |
| InChI Key | AWJUIBRHMBBTKR-UHFFFAOYSA-N |
| Synonym | 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech |
| PubChem CID | 8405 |
| ChEBI | CHEBI:16092 |
| IUPAC Name | isoquinoline |
| SMILES | C1=CC=C2C=NC=CC2=C1 |
Chemical Identifiers
| 119-65-3 | |
| 129.16 | |
| AWJUIBRHMBBTKR-UHFFFAOYSA-N | |
| 8405 | |
| isoquinoline |
| C9H7N | |
| MFCD00006898,MFCD31699977 | |
| 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech | |
| CHEBI:16092 | |
| C1=CC=C2C=NC=CC2=C1 |
Specifications
| 119-65-3 | |
| 242°C | |
| MFCD00006898,MFCD31699977 | |
| 2811 | |
| AWJUIBRHMBBTKR-UHFFFAOYSA-N | |
| isoquinoline | |
| 8405 | |
| 129.16 | |
| Crystalline Lumps |
| White-Yellow | |
| C9H7N | |
| 25 g | |
| 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech | |
| C1=CC=C2C=NC=CC2=C1 | |
| 129.16 | |
| CHEBI:16092 | |
| ≥95.0% (GC) | |
| Isoquinoline |
Safety and Handling
EINECSNumber : (5)-3758
RTECSNumber : NW6825000
TSCA : Yes