missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline 95.0+%, TCI America™

Catalog No. I0182500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0182500G 500 g
I018225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0182500G Supplier TCI America Supplier No. I0182500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119-65-3
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006898,MFCD31699977
InChI Key AWJUIBRHMBBTKR-UHFFFAOYSA-N
Synonym 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech
PubChem CID 8405
ChEBI CHEBI:16092
IUPAC Name isoquinoline
SMILES C1=CC=C2C=NC=CC2=C1

Specifications

Color White-Yellow
Boiling Point 242°C
Quantity 500 g
UN Number 2811
Formula Weight 129.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Isoquinoline
EINECSNumber (5)-3758
RTECSNumber NW6825000
TSCA Yes