missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline, 97%

Catalog No. AC325031000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325031000 100 g Glass bottle
AC325035000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325031000 Supplier Thermo Scientific Chemicals Supplier No. 325031000

CAS: 119-65-3 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-65-3
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006898,MFCD31699977
InChI Key AWJUIBRHMBBTKR-UHFFFAOYSA-N
Synonym 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech
PubChem CID 8405
ChEBI CHEBI:16092
IUPAC Name isoquinoline
SMILES C1=CC=C2C=NC=CC2=C1

Specifications

Melting Point 24°C to 28°C
Color Brown
Density 1.0900g/mL
Boiling Point 242°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.623
Quantity 100 g
Beilstein 20, 380
Specific Gravity 1.09
Merck Index 15, 5268
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted acids,misible with many organic solvents
Viscosity 3.25 mPa.s
Formula Weight 129.16
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material Isoquinoline, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
Do not eat,drink or smoke when using this product.
IF ON SKIN: Wash with plenty
EINECSNumber 204-341-8
RTECSNumber NW6825000
TSCA TSCA

RUO – Research Use Only