missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoquinoline, 97%
Description
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.
Solubility
Soluble in water (5g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.
Solubility
Soluble in water (5g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 24°C to 27°C |
| Density | 1.09 |
| pH | 7.5 |
| Boiling Point | 241°C to 243°C |
| Flash Point | 107°C (225°F) |
| Odor | Pungent |
| Refractive Index | 1.6245 |
| Quantity | 100 g |
| UN Number | UN2811 |
| Beilstein | 107549 |
| Show More |
Safety and Handling
| Hazard Category | H302-H310-H315-H319 |
| Hazard Statement | GHS H Statement H311-H302-H315-H319 Toxic in contact with skin. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P262-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 204-341-8 |
| RTECSNumber | NW6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only