missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline, 97%

Catalog No. AAB2127922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2127922 100 g
AAB2127936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2127922 Supplier Thermo Scientific Chemicals Supplier No. B2127922

CAS: 119-65-3 | C9H7N | 129.16 g/mol

Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoquinoline is used in the manufacture of dyes, paints, antifungal. It is also used as a solvent for the extraction of resins and terpenes.

Solubility
Soluble in water (5g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 119-65-3
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006898,MFCD31699977
InChI Key AWJUIBRHMBBTKR-UHFFFAOYSA-N
Synonym 2-azanaphthalene, 2-benzazine, isochinolin, benzo c pyridine, beta-quinoline, 2-benzanine, 3,4-benzopyridine, .beta.-quinoline, isochinolin czech
PubChem CID 8405
ChEBI CHEBI:16092
IUPAC Name isoquinoline
SMILES C1=CC=C2C=NC=CC2=C1

Specifications

Melting Point 24°C to 27°C
Density 1.09
pH 7.5
Boiling Point 241°C to 243°C
Flash Point 107°C (225°F)
Odor Pungent
Refractive Index 1.6245
Quantity 100 g
UN Number UN2811
Beilstein 107549
Merck Index 14,5222
Solubility Information Soluble in water (5g/L).
Formula Weight 129.16
Percent Purity ≥97%
Chemical Name or Material Isoquinoline
Show More Show Less
Hazard Category H302-H310-H315-H319
Hazard Statement GHS H Statement
H311-H302-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P262-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 204-341-8
RTECSNumber NW6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only