missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-sulfonic Acid 96.0+%, TCI America™

Catalog No. I047025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I047025G 25 g
I0470250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I047025G Supplier TCI America Supplier No. I047025G

Chemical Identifiers

CAS 27655-40-9
Molecular Formula C9H7NO3S
Molecular Weight (g/mol) 209.219
MDL Number MFCD00134089
InChI Key YFMJTLUPSMCTOQ-UHFFFAOYSA-N
Synonym 5-isoquinolinesulfonic acid, 5-sulfoisoquinoline, 5-isoquinolinesulfonicacid, 5-sulphoisoquinoline, pubchem23533, acmc-1chvq, isoquinoline-5-sulphonic acid, ksc203s9f
PubChem CID 241599
IUPAC Name isoquinoline-5-sulfonic acid
SMILES C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2585
Formula Weight 209.22
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-5-sulfonic Acid
EINECSNumber (5)-0826
TSCA Yes