missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7131667
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
I09421G 1 g
I09425G 5 g
2 options

Catalog No. I09421G

Supplier: TCI America I09421G

Only null left

Chemical Identifiers

CAS 80278-67-7
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD03412483
InChI Key ILRSABOCKMOFGW-UHFFFAOYSA-N
Synonym 5-isoquinolinecarboxaldehyde, isoquinoline-5-carboxaldehyde, 5-formylisoquinoline, ksc447q7n, 5-isoquinolinecarbaldehyde, 5-isoquinolinecarboxaldehyde 6ci,9ci
PubChem CID 7016853
IUPAC Name isoquinoline-5-carbaldehyde
SMILES C1=CC2=C(C=CN=C2)C(=C1)C=O

Specifications

Melting Point 119°C
Color Yellow
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-5-carboxaldehyde