missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7131536
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06715G 5 g
I067125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06715G Supplier TCI America Supplier No. I06715G
Only null left

Chemical Identifiers

CAS 486-73-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00006901
InChI Key XAAKCCMYRKZRAK-UHFFFAOYSA-N
Synonym 1-isoquinolinecarboxylic acid, isoquinolinecarboxylic acid, isoquinoline-1-carboxylicacid, 1-carboxyisoquinoline, isoquinioline-1-carboxylic acid, pubchem7537, acmc-209kds, isoquinoline-1carboxylic acid, ksc170s7d, 1-isoquinoline carboxylic acid
PubChem CID 68092
IUPAC Name isoquinoline-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CN=C2C(=O)O

Specifications

Melting Point 156°C
Color Yellow
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-1-carboxylic Acid