missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-1-carbonitrile, 99%

Catalog No. p-7047549
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1942503 1 g
AAL1942506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1942503 Supplier Thermo Scientific Chemicals Supplier No. L1942503

CAS: 1198-30-7 | C10H6N2 | 154.172 g/mol

Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoquinoline-1-carbonitrile is used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines. It is also used as intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1198-30-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00134166
InChI Key HJHXYSBRTVFEDD-UHFFFAOYSA-N
Synonym 1-cyanoisoquinoline, 1-isoquinolinecarbonitrile, isoquinaldonitrile, isoquinolinecarbonitrile, 1-cyano-isoquinoline, isoquinaldinic nitrile, pubchem6269, maybridge1_002114, timtec-bb sbb002487
PubChem CID 306057
IUPAC Name isoquinoline-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CN=C2C#N

Specifications

Melting Point 87°C to 91°C
Odor Odorless
Quantity 1 g
Beilstein 119104
Solubility Information Insoluble in water.
Formula Weight 154.17
Percent Purity 99%
Chemical Name or Material Isoquinoline-1-carbonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only