missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmalonic acid, 98+%

Catalog No. AAL1017106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1017106 5 g
AAL1017114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1017106 Supplier Thermo Scientific Chemicals Supplier No. L1017106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 601-79-6 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 601-79-6
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00021717
InChI Key DQEUFPARIOFOAI-UHFFFAOYSA-N
Synonym isopropylmalonic acid, 2-isopropylmalonic acid, malonic acid, isopropyl, 2-isopropylpropanedioic acid, propanedioic acid, 1-methylethyl, .alpha.-carboxyisovaleric acid, 2-propan-2-yl propanedioic acid, isopropylmalonsaure, isopropyl malonic acid, iso-propyl malonic acid
PubChem CID 69037
IUPAC Name 2-propan-2-ylpropanedioic acid
SMILES CC(C)C(C(O)=O)C(O)=O

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Beilstein 1723234
Formula Weight 146.14
Percent Purity ≥98%
Chemical Name or Material Isopropylmalonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-008-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only