missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropylidenemalononitrile 98.0+%, TCI America™
$190.13 - $1480.06
Chemical Identifiers
| CAS | 13166-10-4 |
|---|---|
| Molecular Formula | C6H6N2 |
| Molecular Weight (g/mol) | 106.128 |
| MDL Number | MFCD00082572 |
| InChI Key | NVBHWAQBDJEGEO-UHFFFAOYSA-N |
| Synonym | 1,1-Dicyano-2-methyl-1-propene |
| PubChem CID | 533473 |
| IUPAC Name | 2-propan-2-ylidenepropanedinitrile |
| SMILES | CC(=C(C#N)C#N)C |
Chemical Identifiers
| 13166-10-4 | |
| 106.128 | |
| NVBHWAQBDJEGEO-UHFFFAOYSA-N | |
| 533473 | |
| CC(=C(C#N)C#N)C |
| C6H6N2 | |
| MFCD00082572 | |
| 1,1-Dicyano-2-methyl-1-propene | |
| 2-propan-2-ylidenepropanedinitrile |
Specifications
| 13166-10-4 | |
| 112°C | |
| MFCD00082572 | |
| 3276 | |
| NVBHWAQBDJEGEO-UHFFFAOYSA-N | |
| 2-propan-2-ylidenepropanedinitrile | |
| 533473 | |
| ≥98.0% (GC) | |
| Isopropylidenemalononitrile |
| Yellow | |
| C6H6N2 | |
| 25 g | |
| 1,1-Dicyano-2-methyl-1-propene | |
| CC(=C(C#N)C#N)C | |
| 106.128 | |
| 106.13 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator