missing translation for 'onlineSavingsMsg'
Learn More

Isopropyldiphenylphosphine 97.0+%, TCI America™

Catalog No. I05831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05831G 1 g
I05835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05831G Supplier TCI America Supplier No. I05831G
Only null left

Chemical Identifiers

CAS 6372-40-3
Molecular Formula C15H17P
Molecular Weight (g/mol) 228.28
MDL Number MFCD00015028
InChI Key LLZAIAIZAVMQIG-UHFFFAOYSA-N
Synonym isopropyldiphenylphosphine, phosphine, 1-methylethyl diphenyl, isopropyldiphenylphosphane, isopropyl diphenyl phosphine, acmc-209nho, isopropyl-diphenylphosphine, diphenyl propan-2-yl phosphane, i-propyldiphenylphosphine
PubChem CID 80754
IUPAC Name diphenyl(propan-2-yl)phosphane
SMILES CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 40°C
Color White
Quantity 1 g
Formula Weight 228.27
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyldiphenylphosphine