Learn More
Isopropylboronic acid pinacol ester, 97%
CAS: 76347-13-2 | C9H19BO2 | 170.059 g/mol
$48.55 - $529.59
Chemical Identifiers
| CAS | 76347-13-2 |
|---|---|
| Molecular Formula | C9H19BO2 |
| Molecular Weight (g/mol) | 170.059 |
| MDL Number | MFCD05663856 |
| InChI Key | MECCSFDHAVAAAW-UHFFFAOYSA-N |
| Synonym | 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| PubChem CID | 12727711 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3386103
|
Thermo Scientific Chemicals
H3386103 |
1 g |
Each for $48.55
|
|
|||||
|
AAH3386106
|
Thermo Scientific Chemicals
H3386106 |
5 g |
Each for $138.05
|
|
|||||
|
AAH3386114
|
Thermo Scientific Chemicals
H3386114 |
25 g |
Each for $529.59
|
|
|||||
Description
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsopropylboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 76347-13-2 | |
| 170.059 | |
| MECCSFDHAVAAAW-UHFFFAOYSA-N | |
| 12727711 | |
| B1(OC(C(O1)(C)C)(C)C)C(C)C |
| C9H19BO2 | |
| MFCD05663856 | |
| 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane |
Specifications
| 76347-13-2 | |
| C9H19BO2 | |
| MFCD05663856 | |
| 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| MECCSFDHAVAAAW-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane | |
| 12727711 | |
| 98% |
| 115°C (60 mmHg) | |
| 1.409 | |
| 1 g | |
| Slightly soluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C(C)C | |
| 170.059 | |
| 170.06 | |
| Isopropylboronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only