missing translation for 'onlineSavingsMsg'
Learn More

Isopropylboronic acid pinacol ester, 97%

Catalog No. AAH3386103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3386103 1 g
AAH3386106 5 g
AAH3386114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3386103 Supplier Thermo Scientific Chemicals Supplier No. H3386103

CAS: 76347-13-2 | C9H19BO2 | 170.059 g/mol

Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 76347-13-2
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.059
MDL Number MFCD05663856
InChI Key MECCSFDHAVAAAW-UHFFFAOYSA-N
Synonym 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12727711
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(C)C

Specifications

Boiling Point 115°C (60 mmHg)
Refractive Index 1.409
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 170.06
Percent Purity 98%
Chemical Name or Material Isopropylboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only